Analtech TLC Developing Tank with LatchLid™ and Aluminum Rack, Glass, Holds Six 10 x 10 cm Plates
Representative image only.
Analtech Mfr # A70-25 – Item # WW-34105-05
Designed for the development of TLC plates
  • Tanks are constructed of durable glass and racks are constructed of anodized aluminum
PRODUCT OPTIONS
₹72,300.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Backing MaterialGlass
  • Plate Length (cm)10
  • Plate Width (cm)10
  • Plate Capacity6
  • Descriptiondeveloping latch-lid tank with lid and rack, capacity: six 10 x 10 cm plates
Key Features
  • Tanks are constructed of durable glass and racks are constructed of anodized aluminum
More About this Item
Tanks accommodate standard TLC plates. The top surface is ground to a precise tolerance for tight sealing to enable tank saturation and optimal plate development. Thinline™ tanks accommodate two plates and require less solvent for development. Latch-Lid™ units hold lids in place with stainless steel latches. Order rack to develop maximum number of plates in a tank.