Allegro 9245-02 Two Man Full Faceshield Supplied Air System with 50 foot Hose
Representative image only.
Allegro Mfr # 9245-02 – Item # WW-81520-31
Complete and convenient NIOSH-approved systems you can take any where
SPECS & DESCRIPTION
Specifications & Description
  • ComplianceNIOSH; ANSI Z87.1-2003
  • ColorBlack
  • DescriptionTwo Man Full Faceshield Supplied Air System with 50 foot Hose
More About this Item
Complete, continuous-flow low-pressure respirator systems for one or two workers are NIOSH approved. These convenient systems come in one box with one part number for easy ordering. Each system has respirator(s), breathing tube(s), air hose(s) and ambient air pump.
Full-Faceshield Systems give you the convenience of a supplied air system with the protection of an impact-resistant faceshield. High impact polycarbonate lens also features anti-fog and anti-scratch coating. Faceshield features a flip-up nylon visor with a polypropylene chin guard for added strength and durability. Cotton shroud keeps debris from falling down the back of the neck. Faceshield assembly meets ANSI Z87.1-2003 for impact resistance.
Approval: NIOSH (as a system)