Air Impurities Removal Systems Final Filters for Ductless Air Filter Systems, 4/pk
Representative image only.
Air Impurities Removal Systems Mfr # RF-987-6 – Item # WW-33703-49
₹30,400.00INR / Pkg of 4
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionFinal Filters for Ductless Air Filter Systems, 4/pk
More About this Item
Easily install or change filters in just seconds. Filter packages designed to suit your specific working conditions
High-Efficiency Filters captures smoke and dust — ideal if you mix dry chemicals, sand, grind, or do light welding. If you use solvents or paint in the same process, choose a combination high-efficiency filter with refillable 2" thick carbon module. Filter is 95% efficient.
HEPA Filters capture soldering fumes and vapors that contain lead. If you use solvents or paint in the same process, choose a combination HEPA filter with refillable 2" thick carbon module. Filter is 99.97% efficient.
Activated Carbon Filter captures volatile solvents, touch-up paint fumes, and other noxious odors. The primary