Advantech L3-S16 Standard Sieve, 3" dia, Stainless Steel Mesh, 1.18 mm Sieve Opening, No. 16
Representative image only.
Advantech Mfr # L3-S16 – Item # WW-59948-29
Steel mesh meets ASTM and ISO specifications
  • 1 year warranty
PRODUCT OPTIONS
₹25,400.00INR / Each
₹27,000.00 - ₹33,700.00INR / Each
for an additional
₹27,000.00INR / Each
for an additional
₹33,700.00INR / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • MaterialAcrylic frame, stainless steel wire
  • U.S. Standard Screen Size (No.)16
  • Tyler Standard Screen Size (mesh)14
  • Sieve Opening (mm)1.19
  • Inside Diameter (InchShort)3
  • Height (InchShort)0.625
  • DescriptionStandard Sieve, Stainless Steel Mesh, 1.18 mm sieve opening, No. 16
  • Warranty1 Year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Standard sieves with stainless steel mesh meet ASTM and ISO specifications and are supplied with a certificate of compliance. Sieves measure 3" ID and 5/8" H.