Advantec QR10090MM Grade QR100 Quartz Fiber Filters, QR100, 90-mm dia; box of 100.
Representative image only.
Advantec Mfr # QR10090MM – Item # WW-06658-24
For analytical or gravimetric studies
  • 1 year warranty
PRODUCT OPTIONS
₹40,800.00INR / Pkg of 100
SPECS & DESCRIPTION
Specifications & Description
  • MaterialQuartz Microfiber
  • Diameter (mm)90
  • Diameter (cm)9
  • DescriptionGrade QR100 Quartz Fiber Filters, QR100, 90-mm dia; box of 100.
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Quartz fiber filters have very low trace metal content and can withstand temperatures up to 1832°F (1000°C). Use to sample acidic gases at high temperatures or for air pollution analysis. The QR100 Grade filters offer superior chemical resistance and will not absorb acidic gases.