Advantec Petri dish without pads, 50x11 mm, 100/cs
Representative image only.
Advantec Mfr # 800100 – Item # WW-14005-24
No need to presterilize
  • Sterilized dishes are ready to use
  • Use only what you need—come in convenient packs of 20
  • Polystyrene dishes come with or without 47-mm absorbent cellulose pad that holds 1.8 to 2.0 mL of liquid media
  • Ridges for easy stacking
  • 1 year warranty
PRODUCT OPTIONS
₹10,200.00INR / Pkg of 100
Parafilm PM992 M Wrap, 2
₹3,900.00INR / Roll of 1
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolystyrene
  • Cap MaterialPolystyrene (PS)
  • Overall Height (in)7/16
  • Overall Height (cm)1.1
  • Outside Diameter (OD) (in)1 31/32
  • Outside Diameter (OD) (cm)5
  • DescriptionPetri dish without pads, 50x11 mm, 100/cs
  • Warranty1 year
1 Year Warranty
Key Features
  • Sterilized dishes are ready to use
  • Use only what you need—come in convenient packs of 20
  • Polystyrene dishes come with or without 47-mm absorbent cellulose pad that holds 1.8 to 2.0 mL of liquid media
  • Ridges for easy stacking
  • 1 year warranty
More About this Item
Ideal for culturing microorganisms in water/wastewater testing
Standard 50 x 11 mm polystyrene dishes. Individual presterilized pads can also be purchased separately. Manufacturer’s certification of compliance available upon request.