Advantec DP7042.5MM DP70 Glass Fiber Filters, 42.5-mm Diameter; 50/Box
Representative image only.
Advantec Mfr # DP7042.5MM – Item # WW-06658-01
For analytical or gravimetric studies
  • 1 year warranty
PRODUCT OPTIONS
₹2,900.00INR / Pkg of 50
SPECS & DESCRIPTION
Specifications & Description
  • MaterialGlass Microfiber
  • Diameter (mm)42.5
  • Diameter (cm)4.25
  • DescriptionDP70 Glass Fiber Filters, 42.5-mm Diameter; 50/Box
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
High wet strength and very high loading capacity; dust and fine particulate sampling.