This item is now discontinued. We recommend the following as suggested replacements: WW-60060-93
Advantec 370S3000N Air monitoring cassette, 3-piece, 37-mm diameter; pack of 50
Representative image only.
Advantec Mfr # 370S3000N – Item # WW-02935-32
Assemble you own monitor for specific applications
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Filter Diameter (mm)37
  • Body MaterialStyrene Acrylonitrile
  • DescriptionAir monitoring cassette, 3-piece, 37-mm diameter; pack of 50
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Empty cassettes and cellulose pads are available as an option for you to assemble a filter/monitoring unit with an MCE membrane filter for asbestos monitoring or other 37-mm filter.