Advanced Green 3 Low-Bleed Septa, 11/16" (17 mm) AG3 CenterGuide; 50/Pk
Representative image only.
Advanced Green 3 Mfr # 246624 – Item # WW-01994-56
True long-life, high temperature green spectrum
  • Reduced injection port sticking and get more injections per septum
  • 3 month warranty
PRODUCT OPTIONS
₹14,200.00INR / Pkg of 50
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionSepta, 11/16\" (17 mm); 50/Pack
  • Warranty3 month
3 Month Warranty
Key Features
  • Reduced injection port sticking and get more injections per septum
  • 3 month warranty
More About this Item
Advanced Green 3 are low-bleed septa that are specifically created to combine a long injection life with low injection port adhesion to meet even your most challenging analysis. Maximum temperature 305°C. Pre-piercing eliminates injection coring on select CenterGuide septa.