AccuMac 9050 Low Emf Precision Switch
Representative image only.
AccuMac Mfr # 9050 – Item # WW-90452-75
Save time and money with 12 channels
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionLow Emf Precision Switch
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
This Low EMF Precision Switch is designed as a useful tool for calibration of bell ross replica platinum resistance thermometers, thermocouples and other temperature sensors/resistors. It offers up to 12 Channels that allow users to measure and compare multiple probes/sensors seamlessly without changing lead connections. The dial switch is uniquely designed to minimize any thermal offsets.