Accuform ZFD807VP NFPA Notice Chemical Hazard, Sign Only, 14" X 10", plastic
Representative image only.
Accuform Mfr # ZFD807VP – Item # WW-41010-07
Signs and kit to help you with regulatory facility compliance
PRODUCT OPTIONS
₹3,100.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • HeaderNOTICE
  • LanguagesEnglish
  • MaterialPlastic
  • Width (in)10
  • Height (in)14
  • DescriptionNFPA Notice Chemical Hazard, Sign Only, 14\" X 10\", plastic
More About this Item
These signs display the four-color National Fire Protection Association (NFPA) diamond format. Signs feature blank hazard rating panels — write in the information for your specific use.