Accuform MLT406CTP Tag, Danger Do Not Operate (white Block), 5 7/8" x 3 1/8", Cardstock, 25/pk
Representative image only.
Accuform Mfr # MLT406CTP – Item # WW-40503-09
Often bought with…
SPECS & DESCRIPTION
Specifications & Description
  • HeaderDANGER
  • LanguagesEnglish
  • MaterialPolyolefin
  • DescriptionTag, Danger Do Not Operate (white Block), 5 7/8\" x 3 1/8\", Cardstock, 25/pk
More About this Item
Tags comply with tagout requirements of the OSHA Lockout/Tagout standard 29 CFR 1910.147. Use tags to mark and identify the lockout situation to prevent the accidental or unexpected start up or release of stored energy when working on equipment and machinery. Tags feature top gabled corners and 3/8” (9.5-mm) hole. All tags are sold in packages of 25.
Compliance: OSHA 29 CFR 1910.147
Cardstock is an economical, 10-mil polyolefin-based material containing no wood-based pulp paper.