Accuform MGT200CTP Tag, Caution, 5 7/8" X 3 1/8", PF-Cardstock
Representative image only.
Accuform Mfr # MGT200CTP – Item # WW-50503-00
SPECS & DESCRIPTION
Specifications & Description
  • LegendBlank
  • HeaderCAUTION
  • LanguagesEnglish
  • MaterialCardstock
  • DescriptionTag, Caution, 5 7/8\" X 3 1/8\", PF-Cardstock
More About this Item
OSHA/ANSI-Compliant Header Tags meet OSHA 29 CFR 1910.145 (f) and ANSI Z535.5 standards which provide specifications, designs, and guidelines for the use and application of accident prevention tags. These tags clearly indicate a temporary hazard that may lead to personal injury or property damage; use until that hazard is eliminated or hazardous operation is complete.
All tags can be written on and have top gabled corners with 3⁄8'' reinforced metal grommet on vinyl plastic.