Accuform LZH616EV2 GHS Label,Skull/Crossbones Pictogram,2" x 2",adhesive poly,250/roll
Representative image only.
Accuform Mfr # LZH616EV2 – Item # WW-50712-94
₹6,600.00INR / Roll of 250
SPECS & DESCRIPTION
Specifications & Description
  • HeaderSKULL/CROSSBONES
  • LanguagesEnglish
  • MaterialAdhesive Poly
  • Width (in)2
  • Height (in)2
  • Quantity250/roll
  • DescriptionGHS Label,Skull/Crossbones Pictogram,2\" x 2\",adhesive poly,250/roll
More About this Item
GHS (Globally Harmonized System) is a worldwide program for classifying chemicals in the workplace. GHS recognizes nine hazard pictograms. The pictograms identify the hazard which helps users take proper precautions to minimize adverse effects.
GHS Skull and Crossbones pictogram indicates the following potential hazard: acute toxicity (fatal or toxic).