Accuform LZH614EV5 GHS Label, Corrosion Pictogram, 2" x 2", adhesive poly, 500/roll
Representative image only.
Accuform Mfr # LZH614EV5 – Item # WW-50712-79
SPECS & DESCRIPTION
Specifications & Description
  • HeaderCORROSION
  • LanguagesEnglish
  • MaterialAdhesive Poly
  • Width (in)2
  • Height (in)2
  • Quantity500/roll
  • DescriptionGHS Label, Corrosion Pictogram, 2\" x 2\", adhesive poly, 500/roll
More About this Item
GHS (Globally Harmonized System) is a worldwide program for classifying chemicals in the workplace. GHS recognizes nine hazard pictograms. The pictograms identify the hazard which helps users take proper precautions to minimize adverse effects.
GHS Corrosion pictogram indicates the following potential hazard: skin corrosion/burns, eye damage, and corrosive to metals.