Accuform LZH605PS5 GHS Label, Gas Cylinder Pictogram, 1" x 1", adhesive paper, 500/roll
Representative image only.
Accuform Mfr # LZH605PS5 – Item # WW-50712-81
SPECS & DESCRIPTION
Specifications & Description
  • HeaderGAS CYLINDER
  • LanguagesEnglish
  • MaterialAdhesive Paper
  • Width (in)1
  • Height (in)1
  • Quantity500/roll
  • DescriptionGHS Label, Gas Cylinder Pictogram, 1\" x 1\", adhesive paper, 500/roll
More About this Item
GHS (Globally Harmonized System) is a worldwide program for classifying chemicals in the workplace. GHS recognizes nine hazard pictograms. The pictograms identify the hazard which helps users take proper precautions to minimize adverse effects.
GHS Gas Cylinder pictogram indicates the following potential hazard: gases under pressure.