Accuform LZH603PS2 GHS Label,Exploding Bomb Pictogram,1" x 1",adhesive paper,250/roll
Representative image only.
Accuform Mfr # LZH603PS2 – Item # WW-50712-64
SPECS & DESCRIPTION
Specifications & Description
  • HeaderEXPLODING BOMB
  • LanguagesEnglish
  • MaterialAdhesive Paper
  • Width (in)1
  • Height (in)1
  • Quantity250/roll
  • DescriptionGHS Label,Exploding Bomb Pictogram,1\" x 1\",adhesive paper,250/roll
More About this Item
GHS (Globally Harmonized System) is a worldwide program for classifying chemicals in the workplace. GHS recognizes nine hazard pictograms. The pictograms identify the hazard which helps users take proper precautions to minimize adverse effects.
GHS Exploding Bomb pictogram indicates the following potential hazard: explosives, self-reactives, and organic peroxides.