Accuform KDH733 Aluma-Tag Hasp, Danger-Locked Out Do Not Operate, 7x3.1", Aluminum, 1/Pk
Representative image only.
Accuform Mfr # KDH733 – Item # WW-40503-39
SPECS & DESCRIPTION
Specifications & Description
  • HeaderDANGER
  • LanguagesEnglish
  • MaterialCardstock, Laminate
  • DescriptionAluma-Tag Hasp, Danger-Locked Out Do Not Operate, 7x3.1\", Aluminum, 1/Pk
More About this Item
These tags comply with tagout requirements of the OSHA Lockout/Tagout standard 29 CFR 1910.147. Use tags to mark and identify the lockout situation to prevent the accidental or unexpected start up or release of stored energy when working on equipment and machinery. Tags feature top gabled corners and 3/8” hole. Choose from three materials based on application.
Cardstock (PF) is a wood pulp-free, tear-resistant, waterproof material, and is 10-mil poly-based.
Plastic (RV) is a 15-mil rigid vinyl plastic material that is durable and will withstand tears, weathering, and various chemicals.
Laminate (HS) is a tough durable poly-tag material for abusive, dirty, and greasy environments. The poly-tag is sealed on both sides by thick rigid vinyl. The plastic and laminate tags also have grommet r