A&D Weighing Pipette Leak Tester, RS-232, 120/220 VAC
Representative image only.
A&D Weighing Mfr # AD-1690 – Item # WW-07911-50
Test for leaks due to damaged seals, O-rings, pistons, or tip holders in just seconds
  • Ideal for micropipettes but also tests small pumps, containers, and piping
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Height (in)3
  • Width (in)9.125
  • Depth (InchShort)5
  • DescriptionPipette Leak Tester, RS-232, 120/220 VAC
  • Warranty1 year
1 Year WarrantyConformite Europeenne
Key Features
  • Ideal for micropipettes but also tests small pumps, containers, and piping
  • 1 year warranty
More About this Item
Instantly determine whether your pipettes need repair by testing for leaks. Damaged seals, o-rings, pistons, and tip holders compromise the air tightness of the pipettes, leading to inaccurate dispensing. The leak tester depressurizes the pipette, monitors the pressure, and assigns a “pass” or “fail” based on the rate of air infiltration. Test is completed in less than five seconds. Download your results to a PC through the RS-232 interface. The tester includes three attachments for different pipette tip sizes, from 2 to 10,000 µL. Replaceable air filter protects the leak tester from dust when it intakes air (replacements sold separately - model 07911-52). Replacement tube set with adapter is available as model 07911-51. Optional rechargeable batteries are sold separately as model 07911-53.