96-well Polypropylene PCR Microplate, Half Skirt, Clear, Nonsterile; 50/Pk
Representative image only.
PCRmax Item # WW-67103-52
Ultra-thin and consistent wall thickness allows precise thermal transfer for optimal results
  • Manufactured with 99.9% pure virgin polypropylene
  • RNAse/DNAse-free, nonpyrogenic, nonsterile
  • Compatible with sealing film
  • Single notch corner
PRODUCT OPTIONS
₹43,500.00INR / Pkg of 50
SPECS & DESCRIPTION
Specifications & Description
  • Number of Wells96
  • Well Volume (µL)200
  • Well ShapeRound
  • SterileNo
  • Plate TypeHalf skirt
  • Plate ColorClear
  • MaterialPolypropylene
  • Description96-well Polypropylene PCR Microplate, Half Skirt, Clear, Nonsterile; 50/Pk
Key Features
  • Manufactured with 99.9% pure virgin polypropylene
  • RNAse/DNAse-free, nonpyrogenic, nonsterile
  • Compatible with sealing film
  • Single notch corner
More About this Item
PCRmax® microplates are designed to fit real-time PCR thermal cyclers and sequencers. The construction of pure, clear polypropylene allows for full transparency. Microplates are compatible with PCRmax Alpha Cycler instruments.