3M LC8S10C LeadCheck Swabs, Lead Test Kit; 8 Swabs per Pack
Representative image only.
3M Mfr # IHLC8S10C – Item # WW-35200-30
Instantly determine if lead-safe practices are needed
  • Red means lead — quick color change
  • Easy, clean, and contained — swabs are disposable, convenient, and simple to use
  • Instant results using a no-mess swab — down to 600 ppm
  • EPA recognized for painted wood, metal, plastic and dust
  • Comes as a pack with 8 swabs
SPECS & DESCRIPTION
Specifications & Description
  • TestLead
  • Measurement Points>600 ppm
  • Reagent PlatformSwabs
  • Tests Per Kit8
  • DescriptionLeadCheck Swabs, Lead Test Kit; 8 Swabs per Pack
Key Features
  • Red means lead — quick color change
  • Easy, clean, and contained — swabs are disposable, convenient, and simple to use
  • Instant results using a no-mess swab — down to 600 ppm
  • EPA recognized for painted wood, metal, plastic and dust
  • Comes as a pack with 8 swabs
More About this Item
Simple-to-use swabs change color in the presence of lead and can be used on most any surface—plain and painted, hard and semi-porous. Simply squeeze the middle of the swab to release the reagent and rub the surface to be tested. The white swab turns pink or red if lead is present. Ensure your safety and confirm negative results with the included confirmation card. The entire test sequence takes about 30 seconds.