Cole-Parmer 316 Stainless Steel Three-blade Propeller, 1" dia x 5/16" bore dia
Representative image only.
Cole-Parmer Item # WW-04552-40
Keep your overhead mixer running smoothly
  • 2 year warranty
PRODUCT OPTIONS
₹9,300.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Description316 Stainless Steel Three-blade Propeller, 1\" dia x 5/16\" bore dia
  • Warranty2 year
Two year warranty
Key Features
  • 2 year warranty
More About this Item
Update clamps, mounts, shafts and propellers to accommodate new protocols. These replacement parts will keep your overhead mixer running smoothly.
Three-blade propellers are for general-purpose mixing. Use alone or mount several on a single shaft for thorough agitation.