Microscope Camera with Reduction Lens, C-Mount, High-Speed, 10 MP, USB 3.0
Representative image only.
Item # WW-78905-16
Capture images and stream video directly from your microscope
  • Documentation and analysis for a deeper view of your slide
  • Easy to install and ready for immediate use
  • Capture, annotate, measure or evaluate still images and video
  • High-speed digital camera features frame rate of 25 fps at 1792 x 1372 (Multiple Speed Level) and 7.2 fps at 3584 x 2746
PRODUCT OPTIONS
₹77,000.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Resolution10 MP, 3584 x 2748
  • Objectives0.5X reduction lens
  • Image Sensor1/2.2\" Aptina MT9J003 color CMOS
  • OutputUSB 3.0
  • System RequirementsWindows XP/Vista/7/8/10 (32 & 64 bit) and Linux
  • Data TransferUSB 3.0
  • Power5 VDC over USB Port
  • DescriptionMicroscope Camera with Reduction Lens, C-Mount, High-Speed, 10 MP, USB 3.0
  • Warranty1 Year
1 Year Warranty
Includes
digital camera, 0.5X reduction lens, 6' (1.8 m) USB cable, 30 mm and 30.5 mm adapters, CD with software and user's instructions.
Key Features
  • Documentation and analysis for a deeper view of your slide
  • Easy to install and ready for immediate use
  • Capture, annotate, measure or evaluate still images and video
  • High-speed digital camera features frame rate of 25 fps at 1792 x 1372 (Multiple Speed Level) and 7.2 fps at 3584 x 2746
More About this Item
AmScope™ microscope cameras are designed for ease of use and the ability to output images and video. These cameras can produce still image and video capture, still image measurement, and allow you to make notes, shapes, and watermarks with ease.
Cameras are available in a wide range of resolution and connectivity options for applications ranging from education, conservation, pharmaceutical, or other demanding fields. Microscopes sold separately.
Designed using the C-mount standard, the camera with the included reduction lens and two sizing-adapters can fit 23mm, 30mm, and 30.5mm photo ports and ocular ports. The reduction lens increases the field-of-view to better match what's seen through the eyepieces. Using a standard USB 3.0 connection, the camera is easy to install. Drawing power from the USB connection, no additional connections are needed.