Fermentation Process Workflow
Vent Filter
Many biopharma applications require the use of vent filters on tank ventilation. Generally, these filters are sterilized to avoid risk of contamination. The filters also serve as a pressure stabilizer for the tank environment.
View Vent Filter Products
pH Probe
In the lab or on the process floor, pH probes sense changes in the acidity of the media which is critical for cell vitality. All pH probes (excepting ISFET technology) will slowly leak solution into the media as a means of measurement.
View pH Probe Products
Sample Bottle
Tanks, vessels, and bottles store product between the stages of drug production. They are sterilized or involve a sterilized barrier between the vessel and the product (such as a tank liner) to mitigate the risk of contamination. Tank ports are pre-ordered to fit the needs of the application. Sample bottles will include a vent filter to avoid pressurizing the bottle during dispensation and cross-contamination.
View Sample Bottle Products
Vent Filter
Many biopharma applications require the use of vent filters on tank ventilation. Generally, these filters are sterilized to avoid risk of contamination. The filters also serve as a pressure stabilizer for the tank environment.
View Vent Filter Products
pH Probe
Many biopharma applications require the use of vent filters on tank ventilation. Generally, these filters are sterilized to avoid risk of contamination. The filters also serve as a pressure stabilizer for the tank environment.
View pH Probe Products
Sample Bottle
Tanks, vessels, and bottles store product between the stages of drug production. They are sterilized or involve a sterilized barrier between the vessel and the product (such as a tank liner) to mitigate the risk of contamination. Tank ports are pre-ordered to fit the needs of the application. Sample bottles will include a vent filter to avoid pressurizing the bottle during dispensation and cross-contamination.
View Sample Bottle Products