Cole-Parmer Technical Library
Lightnin Mixer Standards- ATEX Marking
Published with permission from Lightnin
Marking of equipment used in potentially explosive atmospheres according to ATEX Directive 91/9/EC
(also known as ATEX 95 and ATEX 100a) and EN 13463 pt 1-8
1
2
3
4
5
6
7
CE mark Ex mark
II
2 G
EEx ck
IIC
T4
The following information is intended as a general summary of the most common aspects of ATEX marking legislation relevant to Lightnin’s products and should be used as a guide only.
3. Classification of equipment
Equipment Group
Equipment Category
Atmosphere
Protection level
Required Protection Performance & Operation
I (Mines)
M1
Methane & Dust
Very High
Safe with two independent faults and must remain operational
II (Above Ground)
1
Gas, Vapour, Mist, Dust
Very High
Two independent means of protection or safe with two separate faults
II (Above Ground)
2
Gas, Vapour, Mist, Dust
High
Safe in normal operation and if common faults occur
II (Above Ground)
3
Gas, Vapour, Mist, Dust
Low
Safe in normal operation
3,4. Conditions in the Potentially Explosive atmosphere
Flammable Material
Temporary behaviour of the flammable material in the potentially explosive atmosphere
IEC Classification of the potentially explosive atmospheres
Required marking of the applied device per ATEX
Equipment group
Equipment category
Gases, vapours
• Are present continuously, for long periods or frequently
Zone 0
II
1G
• Are likely to occur
Zone 1
II
2G
(1G equipment may be used)
• Are unlikely to occur or, if they do occur, are likely to do so only infrequently and for a short period only
Zone 2
II
3G
(1G or 2G equipment may be used)
Dusts
• Are present continuously
Zone 20
II
1D
• Are likely to occur
Zone 21
II
2D
(1D equipment may be used)
• Are unlikely to occur in case of whirled dust or, if they do occur, are likely to do so only infrequently
and for a short period only
Zone 22
II
3D
(1D or 2D equipment may be used)
Methane, dust
-
Mining
I
M1, M2 or M3
5. Methods of Protection
Method of Protection
Marking
Protection Principle
Electrical equipment for gases, vapours & mists according to CENELEC
Flameproof enclosure
EEx d
Contain the explosion & prevent flame propagation
Increased safety
EEx e
No arcs, sparks or hot surfaces
Non Sparking
EEx n
No arcs, sparks or hot surfaces
Intrinsic Safety
EEx i
Limit the energy of the spark and surface temperature
Non-electrical equipment and machinery according to 13463-1 CEN
Constructional Safety
EEx c
Design to prevent ignition sources arising
Liquid immersion
EEx k
Keep the explosive atmosphere out by immersion in fluid
6. Classification of gases and dusts per CENELEC/IEC
Gas Group
Temperature Class
T1
T2
T3
T4
T5
T6
I
Methane
-
-
-
-
-
IIA
Ammoniac
Methane
Ethane
Propane
Ethyl alcohol
Cyclohexane
n-butane
n-hexane
Fuel in general
Aircraft fuel
n-hexane
Fuel oil
Acetaldehyde
-
-
IIB
Lighting gas
Acrylonitrile
Ethylene
Ethylene oxide
Ethylene glycol
Hydrogen sulphide
Ethyl ether
-
-
IIC
Hydrogen
Ethine (Acetylene)
-
-
-
Coal disulphide
7. Temperature Classification
CENELEC/IEC
(Group II) Classification
Max Surface Temperature
Comments
T1
450ºC (842ºF)
Temperature relates to all parts of equipment that can come into contact with the potentially explosive atmosphere
T2
300ºC (572ºF)
T3
200ºC (392ºF)
T4
135ºC (275ºF)
T5
100ºC (212ºF)
T6
85ºC (185ºF)
Tx
Maximum surface temperature not defined
Applied to closed tank units where transmission of temperature from the vessel to mixer parts cannot be controlled by Lightnin. Assessment of machine temperature class is the responsibility of the client.
EXAMPLE EQUIPMENT MARKING
CE mark Ex markII 2 G EEx ck IIC T4
ATEX Category 2 (gas) equipment designated for installation in Zone 1. Protection by constructional safety and liquid immersion, suitable for hydrogen atmosphere with maximum equipment surface temperature of 135ºC.
Related Links: Printable page E-mail a colleague
Complete Selection of Mixers & Stirrers
Complete Selection of Motors
Complete Selection of Pumps
Product Selection Guides  |  Product Maintenance  |  FAQs  |  Technical Library Map
Articles and White Papers  |  Case Studies  |  Conversion Factors and Tech Data