Help
Still have questions?
Live chat by BoldChat
Contact our experts
for FREE product application
service and support.
Call 1-800-323-4340
FREE 2009-2010 General Catalog
Chillers and other Baths and Circulators products are now available to order from the Cole-Parmer Instrument Catalog. Click any Chillers category to view all models and products. Request yours today.
Featured Products
Thermo Scientific NESLAB ThermoFlex Recirculating Chillers
NESLAB ThermoFlex Recirculating Chillers
  • Continuous cooling capacity upto 5000 W for consistent temperature control
  • Reliable, easy-to-use chillers - optimized for most demanding applications
  • Select from models with external or internal pressure regulators, low-flow (PD1) or high-flow (PD2) pumps
from $4594.00 More details
Thermo Scientific NESLAB Merlin Recirculating Chillers
NESLAB Merlin Recirculating Chillers
  • Translucent reservoir lets you check liquid level and fluid condition at a glance
  • Digital display resolution can be set to 1°C or 0.1°C
  • Removable condenser vent allows easy access for routine cleaning
from $6748.00 More details
Chilling Solutions
Cole-Parmer India offers an extensive range of chillers for use in temperature control of laboratory and industrial applications. Cole-Parmer chillers offer:
  • Temperature control from -15°C to +40°C
  • High temperature stability to ±0.1°C
  • Wide choice of bath capacity till 15L
For all your chilling needs requiring high stability with a wide variety of temperature control, select from our range of compact chillers, flow-through refrigerated chillers, refrigerated chillers with immersion probe, immersion coolers, temperature-controlled recirculators and recirculating chillers.
Choose from major brands including;
  • Cole-Parmer® Polystat® chillers
  • Thermo Scientific HAAKE immersion coolers, temperature control systems
  • Thermo Scientific NESLAB immersion coolers, ThermoFlex Recirculating Chillers, Merlin Recirculating Chillers
  • PolyScience® Circulators
Use Cole-Parmer's guide to bath and circulator terminology to determine the best unit for your application and our selection guide to choosing a chiller. Alternatively contact our Application Specialists for free technical support and let us bring over 50 years of temperature control experience to help solve your application needs.
Cole-Parmer India
403-404, B-Wing, Delphi
Hiranandani Business Park, Powai
Mumbai 400 076, India
Tel: 91-22-6716-2222
Fax: 91-22-6716-2211
E-Mail: info@Coleparmer.in
Copyright © 2011
Cole-Parmer Sites  Feedback
Cole-Parmer: 
United States
Canada
China
United Kingdom
India
McAfee? Secure sites help keep you safe from identity theft, credit card fraud, spyware, spam, viruses and online scams