Value Plastics Luer to Threaded Fitting, Straight Adapter, White Nylon, Male Luer Integral Lock Ring 9/16" Hex x 1/4-18 NPT(M) Thread; 25/Pk
Representative image only.
Value Plastics Luer to Threaded Fitting, Straight Adapter, White Nylon, Male Luer Integral Lock Ring 9/16" Hex x 1/4-18 NPT(M) Thread; 25/Pk
Value Plastics | Mfr: 14MTLL-1| Item: 5011993
Prevent leaks with integral lock ring mechanism
₹6,100.00INR / Pkg of 25
₹6,100.00INR / Pkg of 25
Buy It Together
Current Item
Value Plastics Luer to Threaded Fitting, Straight Adapter, White Nylon, Male Luer Integral Lock Ring 9/16" Hex x 1/4-18 NPT(M) Thread; 25/Pk
Value Plastics Luer to Threaded Fitting, Straight Adapter, White Nylon, Male Luer Integral Lock Ring 9/16" Hex x 1/4-18 NPT(M) Thread; 25/Pk
5011993
₹6,100.00INR / Pkg of 25
+ +
Cole-Parmer Soft TPE Tubing, Clear, 1/2" ID x 3/4" OD; 50 Ft
5011656
₹51,900.00INR / Pkg of 1
=
Total Price 58400.00 ADD 3 ITEMS TO CART
Product Description
Key Features
  • Easily transition from a luer to thread fitting
  • Luer fittings allow for a simple yet secure twist-lock connection
  • NPT thread fittings are designed to create a tight seal under pressure
  • Constructed of durable chemical resistance nylon
More About this Item
Value Plastics Male Luer Integral Lock Ring to NPT Thread Adapters are designed to easily transition your fluid transfer system from a luer to thread connection. Luer fittings ensure a tight and reliable connection while allowing for quick and easy assembly and disassembly without the need for tools. The integral lock ring is a feature built into the male luer fitting that ensures the fittings remain firmly connected until intentionally disconnected. NPT fittings are commonly used for sealing and connecting pipe fittings and are designed to create a tight seal when assembled, preventing leakage of fluids or gases under pressure. These adapters are available in various materials to accommodate various applications and compliance requirements.
White nylon is suitable for Gamma radiation, ethylene oxide (EtO), and autoclave sterilization.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Certificate of Compliance
Specifications
  • Fitting MaterialNylon
  • Width (InchShort)9/16
  • Height (InchShort)29/32
  • Connection TypeMale Luer Integral Lock Ring 9/16" Hex x 1/4-18 NPT(M)
  • Number Of Ports2
  • Thread Size NPT(M) (Port 2) (InchShort)1/4
  • Flow PatternStraight
  • Fitting Width (in)9/16
  • Regulatory ApprovalsNon-DEHP;Non-Phthalate;RoHS
  • Luer Connection Type (Port 1)Male Luer Integral Lock Ring 9/16" Hex
  • Luer Connection Type (Port 2)1/4-18 NPT(M) Thread
  • DescriptionLuer to Threaded Fitting, Straight Adapter, White Nylon, Male Luer Integral Lock Ring 9/16" Hex x 1/4-18 NPT(M) Thread; 25/Pk