InnoCal NIST-Traceable Calibration; Calibration Weight/Mass, 101 g to 1 kg; ASTM 4-7; NIST F; OIML M1, M2, M3
Representative image only.
InnoCal NIST-Traceable Calibration; Calibration Weight/Mass, 101 g to 1 kg; ASTM 4-7; NIST F; OIML M1, M2, M3
InnoCal | Item: 1710836
ISO/IEC 17025:2005-Accredited Calibration Report
₹18,700.00INR / Each
₹18,700.00INR / Each
Product Description
Key Features
  • Includes a NIST-traceable calibration certificate that establishes traceability to NIST
  • Certificate lists the environmental conditions present at time of calibration
More About this Item
Mass calibrations are performed by an ISO 17025 accredited sub-contractor. InnoCal, our internal ISO 17025 accredited laboratory manages this sub-contracted service for our customers ensuring that the strictest quality requirements are being maintained. Calibration reports are issued directly to the customer after incoming inspection is performed by InnoCal's metrology staff. All measurements reported are traceable to SI units.
Specifications
  • DescriptionNIST Calibration Service for Masses. 101g to 1kg; ASTM 4-7; NIST F; Oiml M1, M2, M3