Material Safety Data Sheet
Rhodium (III) Chloride, Anhydrous

ACC# 16714
Section 1 - Chemical Product and Company Identification
MSDS Name: Rhodium (III) Chloride, Anhydrous
Catalog Numbers: AC419100000, AC419105000
Synonyms:
Rhodium Trichloride.
Company Identification:

              Acros Organics N.V.
              One Reagent Lane
              Fair Lawn, NJ 07410
For information in North America, call: 800-ACROS-01
For emergencies in the US, call CHEMTREC: 800-424-9300

Section 2 - Composition, Information on Ingredients

CAS# Chemical Name Percent EINECS/ELINCS
10049-07-7 Rhodium Trichloride ca 100 233-165-4

Section 3 - Hazards Identification

EMERGENCY OVERVIEW

Appearance: red-brown to black crystalline powder.
Warning! Causes respiratory tract irritation. Irritant. Causes eye and skin irritation. May be harmful if swallowed. May cause digestive tract irritation. Hygroscopic (absorbs moisture from the air). The toxicological properties of this material have not been fully investigated.
Target Organs: Respiratory system, eyes, skin.


Potential Health Effects
Eye: Causes eye irritation. May cause chemical conjunctivitis.
Skin: Causes skin irritation. May be harmful if absorbed through the skin. May cause contact dermatitis.
Ingestion: May cause gastrointestinal irritation with nausea, vomiting and diarrhea. May be harmful if swallowed.
Inhalation: Causes respiratory tract irritation. The toxicological properties of this substance have not been fully investigated. Can produce delayed pulmonary edema.
Chronic: Adverse reproductive effects have been reported in animals.

Section 4 - First Aid Measures


Eyes: Immediately flush eyes with plenty of water for at least 15 minutes, occasionally lifting the upper and lower eyelids. Get medical aid.
Skin: Get medical aid. Flush skin with plenty of water for at least 15 minutes while removing contaminated clothing and shoes. Wash clothing before reuse.
Ingestion: Never give anything by mouth to an unconscious person. Get medical aid. Do NOT induce vomiting. If conscious and alert, rinse mouth and drink 2-4 cupfuls of milk or water.
Inhalation: Remove from exposure and move to fresh air immediately. If not breathing, give artificial respiration. If breathing is difficult, give oxygen. Get medical aid. Do NOT use mouth-to-mouth resuscitation.
Notes to Physician: Treat symptomatically and supportively.

Section 5 - Fire Fighting Measures


General Information: As in any fire, wear a self-contained breathing apparatus in pressure-demand, MSHA/NIOSH (approved or equivalent), and full protective gear. During a fire, irritating and highly toxic gases may be generated by thermal decomposition or combustion.
Extinguishing Media: Use water spray, dry chemical, carbon dioxide, or appropriate foam.
Flash Point: Not applicable.
Autoignition Temperature: Not applicable.
Explosion Limits, Lower:Not available.
Upper: Not available.
NFPA Rating: (estimated) Health: 2; Flammability: 0; Instability: 0

Section 6 - Accidental Release Measures


General Information: Use proper personal protective equipment as indicated in Section 8.
Spills/Leaks: Vacuum or sweep up material and place into a suitable disposal container. Clean up spills immediately, observing precautions in the Protective Equipment section. Avoid generating dusty conditions. Provide ventilation.

Section 7 - Handling and Storage


Handling: Wash thoroughly after handling. Remove contaminated clothing and wash before reuse. Use with adequate ventilation. Minimize dust generation and accumulation. Avoid contact with eyes, skin, and clothing. Keep container tightly closed. Avoid ingestion and inhalation.
Storage: Store in a tightly closed container. Store in a cool, dry, well-ventilated area away from incompatible substances. Store protected from moisture.

Section 8 - Exposure Controls, Personal Protection


Engineering Controls: Facilities storing or utilizing this material should be equipped with an eyewash facility and a safety shower. Use adequate ventilation to keep airborne concentrations low.
Exposure Limits
Chemical Name ACGIH NIOSH OSHA - Final PELs
Rhodium Trichloride 1 mg/m3 TWA (listed under Rhodium). 0.1 mg/m3 TWA (fume) (listed under Rhodium).0.001 mg/m3 TWA (as Rh) (listed under Rhodium soluble compounds).100 mg/m3 IDLH (fume) (listed under Rhodium).2 mg/m3 IDLH (as Rh) (listed under Rhodium soluble compounds). 0.1 mg/m3 TWA (listed under Rhodium).0.001 mg/m3 TWA (as Rh) (listed under Rhodium soluble compounds).

OSHA Vacated PELs: Rhodium Trichloride: No OSHA Vacated PELs are listed for this chemical.
Personal Protective Equipment
Eyes: Wear appropriate protective eyeglasses or chemical safety goggles as described by OSHA's eye and face protection regulations in 29 CFR 1910.133 or European Standard EN166.
Skin: Wear appropriate protective gloves to prevent skin exposure.
Clothing: Wear appropriate protective clothing to prevent skin exposure.
Respirators: A respiratory protection program that meets OSHA's 29 CFR 1910.134 and ANSI Z88.2 requirements or European Standard EN 149 must be followed whenever workplace conditions warrant respirator use.

Section 9 - Physical and Chemical Properties


Physical State: Crystalline powder
Appearance: red-brown to black
Odor: odorless
pH: Not available.
Vapor Pressure: Not available.
Vapor Density: Not available.
Evaporation Rate:Not available.
Viscosity: Not available.
Boiling Point: Not available.
Freezing/Melting Point:Not available.
Decomposition Temperature:Not available.
Solubility: Not available.
Specific Gravity/Density:Not available.
Molecular Formula:Cl3Rh
Molecular Weight:209.26

Section 10 - Stability and Reactivity


Chemical Stability: Stable under normal temperatures and pressures. Hygroscopic: absorbs moisture or water from the air.
Conditions to Avoid: Dust generation, excess heat, exposure to moist air or water.
Incompatibilities with Other Materials: Strong oxidizing agents.
Hazardous Decomposition Products: Hydrogen chloride, rhodium/rhodium oxides.
Hazardous Polymerization: Has not been reported

Section 11 - Toxicological Information


RTECS#:      
CAS# 10049-07-7: VI9275000
LD50/LC50:
CAS# 10049-07-7:
     Oral, rat: LD50 = 1302 mg/kg;
.

Carcinogenicity:
CAS# 10049-07-7: Not listed by ACGIH, IARC, NTP, or CA Prop 65.

Epidemiology: Tumorigenic effects have been reported in experimental animals.
Teratogenicity: No information found
Reproductive Effects: Adverse reproductive effects have occurred in experimental animals.
Mutagenicity: Mutagenic effects have occurred in experimental animals.
Neurotoxicity: No information found
Other Studies:

Section 12 - Ecological Information

No information available.

Section 13 - Disposal Considerations

Chemical waste generators must determine whether a discarded chemical is classified as a hazardous waste. US EPA guidelines for the classification determination are listed in 40 CFR Parts 261.3. Additionally, waste generators must consult state and local hazardous waste regulations to ensure complete and accurate classification.
RCRA P-Series: None listed.
RCRA U-Series: None listed.

Section 14 - Transport Information

US DOT Canada TDG
Shipping Name: DOT regulated - small quantity provisions apply (see 49CFR173.4) No information available.
Hazard Class:
UN Number:
Packing Group:

Section 15 - Regulatory Information

US FEDERAL

TSCA
     CAS# 10049-07-7 is listed on the TSCA inventory.
Health & Safety Reporting List
     None of the chemicals are on the Health & Safety Reporting List.
Chemical Test Rules
     None of the chemicals in this product are under a Chemical Test Rule.
Section 12b
     None of the chemicals are listed under TSCA Section 12b.
TSCA Significant New Use Rule
     None of the chemicals in this material have a SNUR under TSCA.
CERCLA Hazardous Substances and corresponding RQs
     None of the chemicals in this material have an RQ.
SARA Section 302 Extremely Hazardous Substances
     None of the chemicals in this product have a TPQ.
Section 313      No chemicals are reportable under Section 313.
Clean Air Act:
     This material does not contain any hazardous air pollutants.
     This material does not contain any Class 1 Ozone depletors.
     This material does not contain any Class 2 Ozone depletors.
Clean Water Act:
     None of the chemicals in this product are listed as Hazardous Substances under the CWA.
     None of the chemicals in this product are listed as Priority Pollutants under the CWA.
     None of the chemicals in this product are listed as Toxic Pollutants under the CWA.
OSHA:
     None of the chemicals in this product are considered highly hazardous by OSHA.
STATE
     CAS# 10049-07-7 can be found on the following state right to know lists: California, (listed as Rhodium), New Jersey, Pennsylvania, (listed as Rhodium), Minnesota, (listed as Rhodium), Massachusetts.

California Prop 65

California No Significant Risk Level: None of the chemicals in this product are listed.

European/International Regulations

European Labeling in Accordance with EC Directives
Hazard Symbols:
     XI
Risk Phrases:

     R 22 Harmful if swallowed.
     R 36/37/38 Irritating to eyes, respiratory system and skin.

Safety Phrases:

     S 26 In case of contact with eyes, rinse immediately with plenty of
     water and seek medical advice.
     S 37/39 Wear suitable gloves and eye/face protection.

WGK (Water Danger/Protection)

     CAS# 10049-07-7: 0
Canada - DSL/NDSL
     CAS# 10049-07-7 is listed on Canada's DSL List.
Canada - WHMIS
     This product has a WHMIS classification of D2A, D2B.
This product has been classified in accordance with the hazard criteria of the Controlled Products Regulations and the MSDS contains all of the information required by those regulations.
Canadian Ingredient Disclosure List
     CAS# 10049-07-7 is listed on the Canadian Ingredient Disclosure List.

Section 16 - Additional Information

MSDS Creation Date: 5/21/1999
Revision #6 Date: 3/22/2006

The information above is believed to be accurate and represents the best information currently available to us. However, we make no warranty of merchantability or any other warranty, express or implied, with respect to such information, and we assume no liability resulting from its use. Users should make their own investigations to determine the suitability of the information for their particular purposes. In no event shall Fisher be liable for any claims, losses, or damages of any third party or for lost profits or any special, indirect, incidental, consequential or exemplary damages, howsoever arising, even if Fisher has been advised of the possibility of such damages.