Material Safety Data Sheet
Mercury(I) chloride
ACC# 13960
|
Section 1 - Chemical Product and Company Identification |
MSDS Name: Mercury(I) chloride
Catalog Numbers:
M159-100, S80079
Synonyms:
Calomel; Mercurous chloride; Mercury monochloride; Mercury
subchloride.
Company Identification:
Fisher Scientific
1 Reagent Lane
Fair Lawn, NJ 07410
For information, call: 201-796-7100
Emergency Number: 201-796-7100
For CHEMTREC assistance, call: 800-424-9300
For International CHEMTREC assistance, call: 703-527-3887
|
Section 2 - Composition, Information on Ingredients
|
| CAS# |
Chemical Name |
Percent |
EINECS/ELINCS |
| 7546-30-7 |
Mercury(I) chloride |
>99.5 | 231-430-9 |
|
Section 3 - Hazards Identification
|
EMERGENCY OVERVIEW
Appearance: white solid.
Warning! Toxic. Harmful if swallowed. Causes eye,
skin, and respiratory tract irritation. May cause allergic skin
reaction. May be harmful if absorbed through the skin. May cause
central nervous system effects. May cause adverse reproductive
effects. Light sensitive. May cause kidney damage. Moisture
sensitive.
Target Organs: Kidneys, central nervous system,
reproductive system.
Potential Health Effects
Eye:
May cause eye irritation. Contact may cause ulceration of the
conjunctiva and cornea.
Skin:
May cause skin irritation. May be harmful if absorbed through the
skin. May cause an allergic reaction in certain individuals.
May cause a gray discoloration of the skin.
Ingestion:
Harmful if swallowed. May cause kidney failure. May cause severe
digestive tract irritation with abdominal pain, nausea, vomiting and
diarrhea. May cause weakness, fatigue, vascular collapse, and
esophagus damage.
Inhalation:
May cause respiratory tract irritation. Irritation may lead to
chemical pneumonitis and pulmonary edema. May cause kidney damage.
Chronic:
Prolonged or repeated inhalation of dusts may cause neurological
damage. May cause kidney injury. May cause tremors, irritability,
loss of memory and intellect. May also cause Pink disease
characterized by skin, cardiovascular, and neurobehavioral
abnormalities.
Long-term exposure may result in discoloration of the lens and cornea
(mercurialentis). Mercurialentis is a sign of exposure and not of
toxicity. Inorganic Mercury substances such as Mercury (I) chloride
have not been shown to be human teratogens, but should be HANDLED
WITH CAUTION since related Mercury compounds effect fertility in
males and females.
|
Section 4 - First Aid Measures
|
Eyes:
Immediately flush eyes with plenty of water for at least 15 minutes,
occasionally lifting the upper and lower eyelids. Get medical aid.
Skin:
Get medical aid. Flush skin with plenty of water for at least 15
minutes while removing contaminated clothing and shoes. Wash clothing
before reuse.
Ingestion:
Call a poison control center. If swallowed, do not induce vomiting
unless directed to do so by medical personnel. Never give anything by
mouth to an unconscious person. Get medical aid.
Inhalation:
Remove from exposure and move to fresh air immediately. If breathing
is difficult, give oxygen. Get medical aid. Do NOT use mouth-to-mouth
resuscitation. If breathing has ceased apply artificial respiration
using oxygen and a suitable mechanical device such as a bag and a
mask.
Notes to Physician:
Treat symptomatically and supportively.
Antidote:
The use of Dimercaprol or BAL (British Anti-Lewisite) as a chelating
agent should be determined by qualified medical personnel.
|
Section 5 - Fire Fighting Measures
|
General Information:
As in any fire, wear a self-contained breathing apparatus in
pressure-demand, MSHA/NIOSH (approved or equivalent), and full
protective gear. During a fire, irritating and highly toxic gases
may be generated by thermal decomposition or combustion.
Extinguishing Media:
Substance is noncombustible; use agent most appropriate to
extinguish surrounding fire.
Flash Point: Not applicable.
Autoignition Temperature: Not applicable.
Explosion Limits, Lower:Not available.
Upper: Not available.
NFPA Rating:
(estimated) Health: 2; Flammability: 0; Instability: 0
|
Section 6 - Accidental Release Measures
|
General Information: Use proper personal protective equipment as indicated
in Section 8.
Spills/Leaks:
Vacuum or sweep up material and place into a suitable disposal
container. Clean up spills immediately, observing precautions in the
Protective Equipment section. Avoid generating dusty conditions.
Provide ventilation.
|
Section 7 - Handling and Storage
|
Handling:
Wash thoroughly after handling. Remove contaminated clothing and
wash before reuse. Use with adequate ventilation. Minimize dust
generation and accumulation. Do not get on skin and clothing. Do not
ingest or inhale.
Storage:
Keep away from sources of ignition. Do not store in direct sunlight.
Store in a tightly closed container. Store in a cool, dry,
well-ventilated area away from incompatible substances.
|
Section 8 - Exposure Controls, Personal Protection
|
Engineering Controls:
Facilities storing or utilizing this material should be equipped
with an eyewash facility and a safety shower. Use adequate general or
local explosion-proof ventilation to keep airborne levels to
acceptable levels.
Exposure Limits
| Chemical Name |
ACGIH |
NIOSH |
OSHA - Final PELs |
| Mercury(I) chloride
|
0.025 mg/m3 TWA (as Hg) (listed under Mercury inorganic compounds).Skin - potential significant contribution to overall exposure by the cutaneous r oute (listed under Mercury inorganic compounds).
|
0.05 mg/m3 TWA (vapor, except organo alkyls, as Hg) (listed under Mercury compounds).10 mg/m3 IDLH (as Hg, not including organo(alkyl) compounds) (listed under Mercury compounds).
|
0.1 mg/m3 Ceiling (listed under Mercury, aryl and inorganic compounds).
|
OSHA Vacated PELs:
Mercury(I) chloride:
No OSHA Vacated PELs are listed for this chemical.
Personal Protective Equipment
Eyes:
Wear appropriate protective eyeglasses or chemical
safety goggles as described by OSHA's eye and face
protection regulations in 29 CFR 1910.133 or European
Standard EN166.
Skin:
Wear appropriate gloves to prevent skin exposure.
Clothing:
Wear appropriate protective clothing to prevent skin
exposure.
Respirators:
Follow the OSHA respirator regulations found in 29
CFR 1910.134 or European Standard EN 149. Use a
NIOSH/MSHA or European Standard EN 149 approved
respirator if exposure limits are exceeded or if
irritation or other symptoms are experienced.
|
Section 9 - Physical and Chemical Properties
|
Physical State: Solid
Appearance: white
Odor: none reported
pH: Not available.
Vapor Pressure: Not available.
Vapor Density: Not available.
Evaporation Rate:Not available.
Viscosity: Not available.
Boiling Point: Not available.
Freezing/Melting Point:Not available.
Decomposition Temperature:> 752 deg F
Solubility: Insoluble in water.
Specific Gravity/Density: 7.1500
Molecular Formula:Cl2Hg2
Molecular Weight:472.086
|
Section 10 - Stability and Reactivity
|
Chemical Stability:
Stable under normal temperatures and pressures. May decompose when
exposed to light.
Conditions to Avoid:
High temperatures, incompatible materials, light, moisture.
Incompatibilities with Other Materials:
Substance may react with acacia, ammonia, alkali chlorides, bromides,
carbonates, cocaine, copper salts, cyanides, hydroxides, iodine,
iodoform, lead salts, silver salts, soap, sulfates, and sulfites.
Hazardous Decomposition Products:
Hydrogen chloride, mercury/mercury oxides.
Hazardous Polymerization: Has not been reported.
|
Section 11 - Toxicological Information
|
RTECS#:
CAS# 7546-30-7:
OV8750000
LD50/LC50:
Not available.
Carcinogenicity:
CAS# 7546-30-7:
Not listed by ACGIH, IARC, NTP, or CA Prop 65.
Epidemiology:
No information available.
Teratogenicity:
Inorganic Mercury substances have not been shown to be
human teratogens.
Reproductive Effects:
Inorganic mercury substances should be HANDLED WITH CAUTION
since related mercury compounds affect fertility in males
and females.
Mutagenicity:
Mutagenic effects have occurred in experimental animals.
Neurotoxicity:
Neurotoxic effects have occurred in humans.
Other Studies:
|
Section 12 - Ecological Information
|
Ecotoxicity:
No data available.
No information available.
Environmental:
No information reported.
Physical:
No information available.
Other:
None.
|
Section 13 - Disposal Considerations
|
Chemical waste generators must determine whether a discarded chemical is classified
as a hazardous waste.
US EPA guidelines for the classification determination are listed in 40 CFR Parts 261.3.
Additionally, waste generators must consult state and local hazardous waste regulations to
ensure complete and accurate classification.
RCRA P-Series: None listed.
RCRA U-Series: None listed.
|
Section 14 - Transport Information
|
| |
US DOT |
Canada TDG |
| Shipping Name: |
MERCURIC CHLORIDE |
No information available. |
| Hazard Class: |
6.1 |
|
| UN Number: |
UN1624 |
|
| Packing Group: |
II |
|
|
Section 15 - Regulatory Information
|
US FEDERAL
TSCA
CAS# 7546-30-7 is listed on the TSCA inventory.
Health & Safety Reporting List
None of the chemicals are on the Health & Safety Reporting List.
Chemical Test Rules
None of the chemicals in this product are under a Chemical Test Rule.
Section 12b
None of the chemicals are listed under TSCA Section 12b.
TSCA Significant New Use Rule
None of the chemicals in this material have a SNUR under TSCA.
CERCLA Hazardous Substances and corresponding RQs
None of the chemicals in this material have an RQ.
SARA Section 302 Extremely Hazardous Substances
None of the chemicals in this product have a TPQ.
Section 313
This material contains Mercury(I)
chloride (listed as Mercury compounds), >99.5%, (CAS# 7546-30-7)
which is subject to the reporting requirements of Section 313 of
SARA Title III and 40 CFR Part 373.
Clean Air Act:
CAS# 7546-30-7 (listed as Mercury compounds) is listed as a hazardous
air pollutant (HAP).
This material does not contain any Class 1 Ozone depletors.
This material does not contain any Class 2 Ozone depletors.
Clean Water Act:
None of the chemicals in this product
are listed as Hazardous Substances under the CWA.
None of the chemicals in this
product are listed as Priority Pollutants under the CWA.
CAS# 7546-30-7 is listed as a Toxic Pollutant under the Clean Water
Act.
OSHA:
None of the chemicals in this product
are considered highly hazardous by OSHA.
STATE
CAS# 7546-30-7 can be found on the
following state right to know lists: California, (listed as Mercury
compounds), New Jersey, Pennsylvania, (listed as Mercury compounds),
Pennsylvania, (listed as Mercury, aryl and inorganic compounds).
California Prop 65
WARNING: This product contains Mercury(I) chloride, listed as
`Mercury compounds', a chemical known to the state of California to
cause developmental reproductive toxicity.
California No Significant Risk Level:
None of the chemicals in this product are listed.
European/International Regulations
European Labeling in Accordance with EC Directives
Hazard Symbols:
XN N
Risk Phrases:
R 22 Harmful if swallowed.
R 36/37/38 Irritating to eyes, respiratory system and skin.
Safety Phrases:
S 13 Keep away from food, drink and animal feeding stuffs.
S 24/25 Avoid contact with skin and eyes.
S 46 If swallowed, seek medical advice immediately and show this con
tainer or label.
WGK (Water Danger/Protection)
CAS# 7546-30-7: No information available.
Canada - DSL/NDSL
CAS# 7546-30-7 is listed on Canada's DSL List.
Canada - WHMIS
This product has a WHMIS classification of D1B, D2B.
This product has been classified in accordance with the hazard
criteria of the Controlled Products Regulations and the MSDS
contains all of the information required by those regulations.
Canadian Ingredient Disclosure List
CAS# 7546-30-7 is listed on the Canadian Ingredient Disclosure List.
|
Section 16 - Additional Information
|
MSDS Creation Date: 4/29/1999
Revision #4 Date: 1/18/2005
The information above is believed to be accurate and represents the best
information currently available to us. However, we make no warranty of
merchantability or any other warranty, express or implied, with respect to
such information, and we assume no liability resulting from its use. Users
should make their own investigations to determine the suitability of the
information for their particular purposes. In no event shall Fisher be liable
for any claims, losses, or damages of any third party or for lost profits
or any special, indirect, incidental, consequential or exemplary
damages, howsoever arising, even if Fisher has been advised of
the possibility of such damages.